missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Klotho Polyclonal Antibody

Description
A suggested positive control is HepG2 cell lysate. PA5-21078 can be used with blocking peptide PEP-1192.
KLOTHO is the systemic anti-aging hormone within the glycosidase1 superfamily. It encodes a type I membrane protein that is abundant in the kidney and brain. In mice, a deficiency in KLOTHO expression leads to various systemic phenotypes resembling human aging such as arteriosclerosis, osteoporosis, and skin atrophy together with growth retardation, short life-span and infertility. Transgenic mice overexpressing KLOTHO have an extended life span by inhibiting insulin/IGF1 signaling. KLOTHO is involved in the regulation of calcium/phosphorus homeostasis by inhibiting the synthesis of active vitamin D and identified as a potential tumor suppressor.
Specifications
Specifications
| Antigen | Klotho |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | KL |
| Gene Accession No. | O35082, Q9UEF7, Q9Z2Y9 |
| Gene Alias | alpha-kl; HFTC3; Kl; Klotho; Klotho peptide; secreted form of Klotho protein |
| Gene Symbols | KL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?