missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KMT3B/NSD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 624.00 €
Specifications
| Antigen | KMT3B/NSD1 |
|---|---|
| Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18685416
|
Novus Biologicals
NBP2-68968-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18608208
|
Novus Biologicals
NBP2-68968 |
100 μg |
624.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KMT3B/NSD1 Polyclonal antibody specifically detects KMT3B/NSD1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| KMT3B/NSD1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Breast Cancer, Cancer, Cellular Markers, Tumor Suppressors | |
| PBS (pH 7.2) and 40% Glycerol | |
| 64324 | |
| IgG | |
| Protein A purified |
| Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Androgen receptor coactivator 267 kDa protein, Androgen receptor-associated protein of 267 kDa, ARA267STO, DKFZp666C163, EC 2.1.1.43, FLJ10684, FLJ22263, FLJ44628, H3 lysine-36 and H4 lysine-20 specific, KMT3BSotos syndrome, Lysine N-methyltransferase 3B, NR-binding SET domain-containing protein, nuclear receptor binding SET domain protein 1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: DNSVLEIPDAFDRTENMLSMQKNEKIKYSRFAATNTRVKAKQKPLISNSHTDHLMGCTKSAEPGTETSQVNLSDLKASTLVHKPQSDFT | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title