missing translation for 'onlineSavingsMsg'
Learn More

KN-62, MedChemExpress

Product Code. 16478317
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478317 5 mg 5mg
16425628 10 mg 10mg
16411797 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16478317 Supplier Medchem Express Supplier No. HY13290
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress KN-62 is a selective and reversible inhibitor of calmodulin-dependent protein kinase II (CaMK-II) with a Ki of 0.9 μM for rat brain CaMK-II. KN-62 directly binds to the calmodulin binding site of CaMK-II. KN-62 displays noncompetitive antagonism at P2X7 receptors in HEK293 cells, with an IC50 value of approximately 15 nM.

KN-62 is a selective and reversible inhibitor of calmodulin-dependent protein kinase II (CaMK-II) with a Ki of 0.9 μM for rat brain CaMK-II. KN-62 directly binds to the calmodulin binding site of CaMK-II. KN-62 displays noncompetitive antagonism at P2X7 receptors in HEK293 cells, with an IC50 value of approximately 15 nM.

Specifications

Chemical Name or Material KN-62
CAS 127191-97-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C38H35N5O6S2
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (138.53 mM)
SMILES O=C(N1CCN(C2=CC=CC=C2)CC1)[C@H](CC3=CC=C(C=C3)OS(=O)(C4=CC=CC5=C4C=CN=C5)=O)N(C)S(=O)(C6=CC=CC7=C6C=CN=C7)=O
Molecular Weight (g/mol) 721.84
Formula Weight 721.84
Percent Purity 98.27%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.