missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KNCN Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48844-25ul
This item is not returnable.
View return policy
Description
KNCN Polyclonal antibody specifically detects KNCN in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| KNCN | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Kino, Kinocilin, L5 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: KARFNLDHILPTIGSLRIHPHPGADHGEGRSSTNGNKEGARSSLSTVSRTLEKLKPG | |
| 25 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 148930 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur