missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Krebs-Ringer Solution, HEPES-buffered

Product Code. 15428779
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15428779 500 mL 500mL
15438779 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15428779 Supplier Thermo Scientific Chemicals Supplier No. J67795.AP
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Standard physiological buffer that is suitable for various calcium imaging experiments. It is also used as a preserving agent.

Standard physiological buffer that is suitable for various calcium imaging experiments. It is also used as a preserving agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Krebs-Ringer Solution
Physical Form Liquid
Quantity 500 mL
Packaging Plastic bottle
Solubility Information Not miscible or difficult to mix in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.