missing translation for 'onlineSavingsMsg'
Learn More

Ksp-Cadherin Rabbit, Clone: CDH16/1532R, enQuireBio™

Product Code. 15999697
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15999697 20 μg 20µg
15909707 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15999697 Supplier EnquireBio Supplier No. 1014RBM2P0

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Specifications

Antigen Ksp-Cadherin
Applications Immunohistochemistry
Cellular Localization Cell Surface with some cytoplasmic
Classification Monoclonal
Clone CDH16/1532R
Concentration 200μg/mL
Conjugate Unconjugated
Formulation 10mM PBS with or without 0.05% BSA and 0.05% azide.
Gene Accession No. NP_001191674
Gene Symbols CDH16
Host Species Rabbit
Immunogen Recombinant human full-length CDH16 protein
Molecular Weight of Antigen 130kDa
Positive Control Normal kidney or renal cell carcinoma.
Purification Method Protein A/G Purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1014
Content And Storage Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.