missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ KU 0060648
GREENER_CHOICE

Product Code. 30001690
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001690

Brand: Tocris Bioscience™ 4840/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dual DNA-PK and PI 3-K inhibitor

Dual PI 3-K and DNA-PK inhibitor (IC50 values are <0.1, 0.5, 4 and 19 nM for PI 3-Kδ, PI 3-Kβ, PI 3-Kα and DNA-PK respectively). Inhibits proliferation of MCF7 cells in vitro and delays growth of MCF7 xenografts in mice. Also enhances CRISPR-Cas9-mediated homology-directed repair (HDR) efficiency ∽2 to 4-fold, and decreases nonhomologous end-joining (NHEJ) frequency ∽40%; maximum effect seen at 250 nM.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 881375-00-4
Molecular Formula C33H34N4O4S

Specifications

Quantity 10 mg
Formula Weight 582.71
Purity >97%
Molecular Weight (g/mol) 582.719
Chemical Name or Material KU 0060648
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.