missing translation for 'onlineSavingsMsg'
Learn More

KU-55933, MedChemExpress

Product Code. 16485696
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16485696 100 mg 100mg
16467667 5 mg 5mg
16464918 10 mg 10mg
16461167 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16485696 Supplier Medchem Express Supplier No. HY12016
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress KU-55933 is a potent ATM inhibitor with an IC50 and Ki of 12.9 and 2.2 nM, respectively, and is highly selective for ATM as compared to DNA-PK, PI3K/PI4K, ATR and mTOR.

KU-55933 is a potent ATM inhibitor with an IC50 and Ki of 12.9 and 2.2 nM, respectively, and is highly selective for ATM as compared to DNA-PK, PI3K/PI4K, ATR and mTOR.

Specifications

Chemical Name or Material KU-55933
Physical Form Solid
CAS 587871-26-9
Color Earth Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H17NO3S2
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 80 mg/mL (202.28 mM; Need ultrasonic)
SMILES O=C1C=C(OC(C2=C3SC4=C(SC3=CC=C2)C=CC=C4)=C1)N5CCOCC5
Molecular Weight (g/mol) 395.49
Formula Weight 395.49
Percent Purity 99.72%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.