missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KV1.1 (KCNA1) Recombinant Rabbit Monoclonal Antibody (HL3162)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556773
This item is not returnable.
View return policy
Description
KV1.1 (KCNA1) Recombinant Monoclonal Antibody for Western Blot
This gene encodes a voltage-gated delayed potassium channel that is phylogenetically related to the Drosophila Shaker channel. The encoded protein has six putative transmembrane segments, and the loop between S5 and S6 forms the pore and contains the conserved selectivity filter motif. The functional channel is a homotetramer. The N-terminus of the channel is associated with beta subunits that can modify the inactivation properties of the channel as well as affect expression levels. The C-terminus of the channel is complexed to a PDZ domain protein that is responsible for channel targeting. Mutations in this gene have been associated with myokymia with periodic ataxia.
Specifications
| KV1.1 (KCNA1) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P10499, P16388 | |
| Kcna1 | |
| The immunogen used to generate this antibody corresponds to human Kv1.1. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| HL3162 | |
| Unconjugated | |
| Kcna1 | |
| AEMK; AI840627; brain potassium channel protein-1; EA1; HBK1; HUK1; IA; Kca1-1; Kcna; Kcna1; Kcpvd; Kv1.1; MBK1; mceph; megencephaly; MGC126782; MGC138385; MK1; Mk-1; MKI; potassium (K+) channel protein voltage dependent; potassium channel, voltage gated shaker related subfamily A, member 1; potassium voltage gated channel shaker related subfamily member 1; potassium voltage gated channel, shaker related subfamily, member 1; potassium voltage-gated channel subfamily A member 1; potassium voltage-gated channel, shaker-related subfamily, member 1; potassium voltage-gated channel, shaker-related subfamily, member 1 (episodic ataxia with myokymia); RBK1; RBKI; RCK1; Shak; Voltage-gated K(+) channel HuKI; voltage-gated potassium channel HBK1; Voltage-gated potassium channel subunit Kv1.1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 16485, 24520 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction