missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-3-Phenyllactic acid, 98%

Product Code. 11423737
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423737 25 g 25g
11403737 1 g 1g
11413737 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423737 Supplier Thermo Scientific Alfa Aesar Supplier No. A18203.14
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 20312-36-1 | C9H9O3 | 165.17 g/mol

L-(-)-3-Phenyllactic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(-)-3-Phenyllactic acid is used as pharmaceutical intermediate.

Solubility
Solubility in water gives very faint turbidity. 

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20312-36-1
Molecular Formula C9H9O3
Molecular Weight (g/mol) 165.17
MDL Number MFCD00004244
InChI Key VOXXWSYKYCBWHO-QMMMGPOBSA-M
Synonym l---3-phenyllactic acid, s-2-hydroxy-3-phenylpropionic acid, s-2-hydroxy-3-phenylpropanoic acid, l--3-phenyllactic acid, s---3-phenyllactic acid, l-3-phenyllactic acid, d-3-phenyllactic acid, 2s-2-hydroxy-3-phenylpropanoic acid, s-3-phenyllactic acid, s---2-hydroxy-3-phenylpropionic acid
PubChem CID 444718
ChEBI CHEBI:43065
IUPAC Name (2S)-2-hydroxy-3-phenylpropanoic acid
SMILES O[C@@H](CC1=CC=CC=C1)C([O-])=O

Specifications

Melting Point 122°C to 126°C
Quantity 25 g
Beilstein 2209792
Optical Rotation −24° (c=1 in Water)
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material L-(-)-3-Phenyllactic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.