missing translation for 'onlineSavingsMsg'
Learn More

L(-)-3-Phenyllactic acid, 98%

Product Code. 10782885 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10782885 25 g Glass bottle 25g
10113440 1 g Glass bottle 1g
10133440 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10782885 Supplier Thermo Scientific Acros Supplier No. 150920250
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 20312-36-1 | C9H9O3 | 165.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20312-36-1
Molecular Formula C9H9O3
Molecular Weight (g/mol) 165.17
MDL Number MFCD00004244
InChI Key VOXXWSYKYCBWHO-QMMMGPOBSA-M
Synonym l---3-phenyllactic acid, s-2-hydroxy-3-phenylpropionic acid, s-2-hydroxy-3-phenylpropanoic acid, l--3-phenyllactic acid, s---3-phenyllactic acid, l-3-phenyllactic acid, d-3-phenyllactic acid, 2s-2-hydroxy-3-phenylpropanoic acid, s-3-phenyllactic acid, s---2-hydroxy-3-phenylpropionic acid
PubChem CID 444718
ChEBI CHEBI:43065
IUPAC Name (2S)-2-hydroxy-3-phenylpropanoic acid
SMILES O[C@@H](CC1=CC=CC=C1)C([O-])=O

Specifications

Melting Point 121°C to 125°C
Color Beige-Brown to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5CH2CH(OH)CO2H
Quantity 25 g
Specific Rotation Condition −18.50° (24°C c=1,CH3COOH)
Specific Rotation -18.5
Solubility Information Solubility in water: soluble
Formula Weight 166.18
Percent Purity 98%
Physical Form Crystals, Powder or Flakes
Chemical Name or Material L(-)-3-Phenyllactic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.