missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ L-745,870 trihydrochloride
GREENER_CHOICE

Product Code. 30001159
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001159 10 mg 10mg
18044392 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001159 Supplier Tocris Bioscience™ Supplier No. 1002/10
  • ONLINE EXCLUSIVE PRICE
    225.00 €
    212.10 € / 10mg
    Save 12.90 €
    5% Off
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Highly selective D4 antagonist

A highly potent and selective D4 dopamine receptor antagonist; L-745,870 has Ki values of 0.51, 2300 and 960 nM for D4, D3 and D2 subtypes respectively and > 1000-fold selectivity over 5-HT2, D1 and D5 receptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 866021-03-6
Molecular Formula C18H22Cl4N4

Specifications

Quantity 10 mg
Formula Weight 436.21
Purity >99%
Molecular Weight (g/mol) 436.202
Chemical Name or Material L-745, 870 trihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.