missing translation for 'onlineSavingsMsg'
Learn More

L 888607 Racemate, MedChemExpress

Product Code. 16493066
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493066 25 mg 25mg
16487137 5 mg 5mg
16485306 100 mg 100mg
16454418 10 mg 10mg
16430737 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16493066 Supplier Medchem Express Supplier No. HY111271A
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L 888607 Racemate is a selective prostaglandin D2 receptor subtype 1 (DP1) antagonist, with Kis of 132 nM and 17 nM for DP1 and thromboxane A2 receptor (TP), respectively.

L 888607 Racemate is a selective prostaglandin D2 receptor subtype 1 (DP1) antagonist, with Kis of 132 nM and 17 nM for DP1 and thromboxane A2 receptor (TP), respectively.

Specifications

Chemical Name or Material L 888607 Racemate
CAS 1030017-51-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H15ClFNO2S
Quantity 25 mg
Solubility Information DMSO : 6 mg/mL (15.96 mM; Need ultrasonic)
SMILES O=C(O)CC1CCN2C1=C(SC3=CC=C(Cl)C=C3)C4=C2C=C(F)C=C4
Molecular Weight (g/mol) 375.84
Formula Weight 375.84
Percent Purity 99.44%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.