missing translation for 'onlineSavingsMsg'
Learn More

L-Adrenaline, 98+%

Product Code. p-7044157
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11381757

Brand: Thermo Scientific Alfa Aesar L04911.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51-43-4 | C9H13NO3 | 183.207 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-43-4
Molecular Formula C9H13NO3
Molecular Weight (g/mol) 183.207
MDL Number MFCD00002204
InChI Key UCTWMZQNUQWSLP-VIFPVBQESA-N
Synonym epinephrine, l-adrenaline, adrenaline, l-epinephrine, adrenalin, levoepinephrine, epinefrina, --adrenaline, adnephrine, chelafrin
PubChem CID 5816
ChEBI CHEBI:28918
IUPAC Name 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
SMILES CNCC(C1=CC(=C(C=C1)O)O)O

Specifications

Melting Point 216°C to 219°C
Quantity 5 g
UN Number UN2811
Beilstein 2368277
Sensitivity Air and light sensitive
Merck Index 14,3619
Optical Rotation −52° (c=1 in 0.6N HCl)
Formula Weight 183.2
Percent Purity ≥98%
Chemical Name or Material L-Adrenaline
compliance-icons compliance-icons
Product Identifier
  • L-Adrenaline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Epinephrine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.