missing translation for 'onlineSavingsMsg'
Learn More

L-alpha-Aminoxy-beta-phenylpropionic Acid Hydrobromide, TRC

Product Code. 30133619
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133619 250 mg 250mg
30005402 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133619 Supplier Toronto Research Chemicals Supplier No. TRCA632500250MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73086-97-2
Molecular Formula C9 H11 N O3 . Br H
Molecular Weight (g/mol) 262.1
Synonym Benzenepropanoic acid, α-(aminooxy)-, hydrobromide, (S)- (9CI), (2S)-2-(Aminooxy)-3-phenylpropanoic acid hydrobromide, (2S)-2-Aminooxy-3-phenylpropanoic acid hydrobromide, L-α-Aminoxy-β-phenylpropionic acid hydrobromide, (αS)-α-(Aminooxy)benzenepropanoic acid hydrobromide
IUPAC Name (2S)-2-aminooxy-3-phenylpropanoic acid;hydrobromide
SMILES Br.NO[C@@H](Cc1ccccc1)C(=O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C9H11NO3.BrH/c10-13-8(9(11)12)6-7-4-2-1-3-5-7;/h1-5,8H,6,10H2,(H,11,12);1H/t8-;/m0./s1
Formula Weight 261.0001
Chemical Name or Material L-alpha-Aminoxy-beta-phenylpropionic Acid, Hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.