missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteine ethyl ester hydrochloride, 98+%

Product Code. 11301677
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11301677 25 g 25g
11311677 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11301677 Supplier Thermo Scientific Alfa Aesar Supplier No. L04692.14
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 868-59-7 | C5H12ClNO2S | 185.666 g/mol

L-Cysteine ethyl ester hydrochloride is widely used in food additive, cosmetic, pharmaceutical area.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Cysteine ethyl ester hydrochloride is widely used in food additive, cosmetic, pharmaceutical area.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from moisture and oxidizing agents. Incompatible with oxidizing agents and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 868-59-7
Molecular Formula C5H12ClNO2S
Molecular Weight (g/mol) 185.666
MDL Number MFCD00012631
InChI Key JFKJWWJOCJHMGV-WCCKRBBISA-N
Synonym l-cysteine ethyl ester hydrochloride, h-cys-oet.hcl, r-ethyl 2-amino-3-mercaptopropanoate hydrochloride, l-cystein ethyl ester hydrochloride, ethyl l-cysteinate hydrochloride, cysteine ethyl ester hydrochloride, l-cysteine ethyl ester hcl, unii-am467tb2i1, ethyl cysteinate hydrochloride, cysteine, ethyl ester, hydrochloride
PubChem CID 2723617
IUPAC Name ethyl (2R)-2-amino-3-sulfanylpropanoate;hydrochloride
SMILES CCOC(=O)C(CS)N.Cl

Specifications

Melting Point 123°C to 125°C
Beilstein 3562600
Solubility Information Soluble in water.
Quantity 25 g
Formula Weight 185.67
Optical Rotation −12° (c=1 in 1N HCl)
Sensitivity Hygroscopic
Percent Purity ≥98%
Chemical Name or Material L-Cysteine ethyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.