missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Epinephrine, 99%

Product Code. 10434253 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10434253 5 g Glass bottle 5g
10219883 1 g Glass bottle 1g
10740965 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10434253 Supplier Thermo Scientific Acros Supplier No. 204400050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 51-43-4 | C9H13NO3 | 183.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-43-4
MDL Number MFCD00002204
InChI Key UCTWMZQNUQWSLP-VIFPVBQESA-N
Synonym epinephrine, l-adrenaline, adrenaline, l-epinephrine, adrenalin, levoepinephrine, epinefrina, --adrenaline, adnephrine, chelafrin
PubChem CID 5816
ChEBI CHEBI:28918
IUPAC Name 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
SMILES CNCC(C1=CC(=C(C=C1)O)O)O

Specifications

Melting Point 208.0°C to 211.0°C
Color White to Beige
Loss on Drying 1% max. (18 hrs) (0.7 kPa, P2O5)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C9H13NO3
Linear Formula (HO)2C6H3CH(CH2NHCH3)OH
Quantity 5 g
Beilstein 13, 830
Merck Index 15, 3674
Specific Rotation Condition − 51.50 (20.00°C c=4,1M HCl,dry sub)
Specific Rotation − 51.50
Solubility Information Solubility in water: sparingly soluble. Other solubilities: insoluble in alcohol, chloroform, ether and, acetone
Molecular Weight (g/mol) 183.21
Formula Weight 183.21
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material L(-)-Epinephrine, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.