missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic Acid-L-glycine-L-glycine-L-isoleucine-L-tryptophan-L-phenylalanine-L-proline-L-tyrosine-L-glycine-L-glycine-L-phenylalanine TFA Salt (95%), TRC

Product Code. 30403359
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30403359 25 mg 25mg
30396128 10 mg 10mg
30394736 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30403359 Supplier Toronto Research Chemicals Supplier No. TRCG14710525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Glutamic Acid-L-glycine-L-glycine-L-isoleucine-L-tryptophan-L-phenylalanine-L-proline-L-tyrosine-L-glycine-L-glycine-L-phenylalanine TFA Salt
Analyte or Component Names L-Glutamic Acid-L-glycine-L-glycine-L-isoleucine-L-tryptophan-L-phenylalanine-L-proline-L-tyrosine-L-glycine-L-glycine-L-phenylalanine TFA Salt
Shipping Condition Room Temperature
Molecular Formula C62H76N12O15 •(C2HF3O2)
Synonym 5-((1H-Indol-3-yl)methyl)-17-amino-2-benzyl-8-(sec-butyl)-1-(2-((1-((2-((2-((1-carboxy-2-phenylethyl)amino)-2-oxoethyl)amino)-2-oxoethyl)amino)-3-(4-hydroxyphenyl)-1-oxopropan-2-yl)carbamoyl)pyrrolidin-1-yl)-1,4,7,10,13,16-hexaoxo-3,6,9,12,15-pentaazaicosan-20-oic Acid Trifluoroacetic Acid Salt
InChI Formula InChI=1S/C62H76N12O15.C2HF3O/c1-3-37(2)56(72-54(80)34-65-52(78)35-67-73-42(36-75)22-25-55(81)82)60(86)70-47(30-41-31-63-45-18-11-10-17-44(41)45)58(84)71-48(28-38-13-6-4-7-14-38)61(87)74-26-12-19-50(74)59(85)69-46(27-40-20-23-43(76)24-21-40)57(83)66-32-51(77)64-33-53(79)68-49(62(88)89)29-39-15-8-5-9-16-39;3-2(4,5)1-6/h4-11,13-18,20-21,23-24,31,36-37,42,46-50,56,63,67,73,76H,3,12,19,22,25-30,32-35H2,1-2H3,(H,64,77)(H,65,78)(H,66,83)(H,68,79)(H,69,85)(H,70,86)(H,71,84)(H,72,80)(H,81,82)(H,88,89);1H/t37-,42-,46-,47-,48-,49-,50-,56-;/m0./s1
Solution Type Neat
SMILES OC([C@H](CC1=CC=CC=C1)NC(CNC(CNC([C@H](CC2=CC=C(C=C2)O)NC([C@H]3N(C([C@H](CC4=CC=CC=C4)NC([C@H](CC5=CNC6=C5C=CC=C6)NC([C@H]([C@H](CC)C)NC(CNC(CNN[C@H](C([H])=O)CCC(O)=O)=O)=O)=O)=O)=O)CCC3)=O)=O)=O)=O)=O.O=CC(F)(F)F
IUPAC Name (2S,5S,8S,17S)-5-((1H-indol-3-yl)methyl)-2-benzyl-8-((S)-sec-butyl)-1-((S)-2-(((S)-1-((2-((2-(((S)-1-carboxy-2-phenylethyl)amino)-2-oxoethyl)amino)-2-oxoethyl)amino)-3-(4-hydroxyphenyl)-1-oxopropan-2-yl)carbamoyl)pyrrolidin-1-yl)-17-formyl-1,4,7,10,13-pentaoxo-3,6,9,12,15,16-hexaazaicosan-20-oic acid 2,2,2-trifluoro-1l3-ethan-1-one
Industry Type Pharmaceutical
Molecular Weight (g/mol) 1327.4
Quantity 25 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.