missing translation for 'onlineSavingsMsg'
Learn More

L-Lactic acid, MedChemExpress

Product Code. 16453469
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16453469 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16453469 Supplier Medchem Express Supplier No. HYY0479
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L-Lactic acid is a buildiing block which can be used as a precursor for the production of the bioplastic polymer poly-lactic acid.

L-Lactic acid is a buildiing block which can be used as a precursor for the production of the bioplastic polymer poly-lactic acid.

Specifications

Chemical Name or Material L-Lactic acid
Physical Form Solid
CAS 79-33-4
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C3H6O3
For Use With (Application) COVID-19-immunoregulation
Synonym (S)-2-Hydroxypropanoic acid
Solubility Information H2O : 100 mg/mL (1110.12 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (1110.12 mM; Need ultrasonic)
SMILES C[C@H](O)C(O)=O
Molecular Weight (g/mol) 90.08
Formula Weight 90.08
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.