missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine methyl ester dihydrochloride, 98%

Product Code. 15500614 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15500614 5 g 5g
15510614 25 g 25g
15520614 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15500614 Supplier Thermo Scientific Acros Supplier No. 459270050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 26348-70-9 | C7H18Cl2N2O2 | 233.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26348-70-9
Molecular Formula C7H18Cl2N2O2
Molecular Weight (g/mol) 233.13
MDL Number MFCD00039067
InChI Key SXZCBVCQHOJXDR-ILKKLZGPSA-N
Synonym l-lysine methyl ester dihydrochloride, h-lys-ome.2hcl, methyl l-lysinate dihydrochloride, l-lysine methyl ester 2hcl, s-methyl 2,6-diaminohexanoate dihydrochloride, l-lysine, methyl ester, dihydrochloride, methyl l-lysinate hcl, lysine, methyl ester, dihydrochloride, methyl 2s-2,6-diaminohexanoate dihydrochloride, h-lys-ome
PubChem CID 117778
IUPAC Name methyl (2S)-2,6-diaminohexanoate;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].COC(=O)[C@@H](N)CCCCN

Specifications

Melting Point 200.0°C
Infrared Spectrum Authentic
Solubility Information Solubility in water: soluble.
Quantity 5 g
Formula Weight 233.14
Percent Purity 98%
Chemical Name or Material L-Lysine methyl ester dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.