missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Lysine methyl ester dihydrochloride, 99%

Product Code. 11433677
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433677 100 g 100g
11423677 25 g 25g
11443677 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433677 Supplier Thermo Scientific Chemicals Supplier No. A18157.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 26348-70-9 | C7H18Cl2N2O2 | 233.13 g/mol

L-Lysine methyl ester dihydrochloride is used in the production of lysine methyl ester based cationic surfactants and hydrogels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Lysine methyl ester dihydrochloride is used in the production of lysine methyl ester based cationic surfactants and hydrogels.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26348-70-9
Molecular Formula C7H18Cl2N2O2
Molecular Weight (g/mol) 233.13
MDL Number MFCD00039067
InChI Key SXZCBVCQHOJXDR-ILKKLZGPSA-N
Synonym l-lysine methyl ester dihydrochloride, h-lys-ome.2hcl, methyl l-lysinate dihydrochloride, l-lysine methyl ester 2hcl, s-methyl 2,6-diaminohexanoate dihydrochloride, l-lysine, methyl ester, dihydrochloride, methyl l-lysinate hcl, lysine, methyl ester, dihydrochloride, methyl 2s-2,6-diaminohexanoate dihydrochloride, h-lys-ome
PubChem CID 117778
IUPAC Name methyl (2S)-2,6-diaminohexanoate;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].COC(=O)[C@@H](N)CCCCN

Specifications

Melting Point ∼200°C (decomposition)
Beilstein 5302183
Solubility Information Soluble in water.
Quantity 100 g
Formula Weight 233.14
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material L-Lysine methyl ester dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.