missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(-)-Menthol, 99.5%

Product Code. 10631063 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10631063 5 g Glass bottle 5g
10467320 100 g Glass bottle 100g
10103370 500 g Glass bottle 500g
10389223 2.5 kg Plastic Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10631063 Supplier Thermo Scientific Chemicals Supplier No. 125400050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2216-51-5 | C10H20O | 156.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2216-51-5
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
MDL Number MFCD00062979
InChI Key NOOLISFMXDJSKH-KXUCPTDWSA-N
Synonym l-menthol, --menthol, levomenthol, menthomenthol, l---menthol, hexahydrothymol, menthacamphor, peppermint camphor, 1r,2s,5r-2-isopropyl-5-methylcyclohexanol, u.s.p. menthol
PubChem CID 16666
ChEBI CHEBI:15409
IUPAC Name (1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-ol
SMILES CC1CCC(C(C1)O)C(C)C

Specifications

Melting Point 41.0°C to 43.0°C
Color Colorless to White
Density 0.8900g/mL
Boiling Point 212.0°C
Flash Point 94°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Packaging Glass bottle
Refractive Index 1.46
Quantity 5 g
Beilstein 06,28
Fieser 13,172; 16,203
Merck Index 15,5905
Specific Gravity 0.89
Specific Rotation Condition −51.00 (20.00°C 589nm,c=10,EtOH)
Specific Rotation − 51.00
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in acetic acid,soluble in alcohol,chloroform,ether,,petroleum ether
Formula Weight 156.27
Percent Purity 99.5%
Physical Form Crystals or Crystalline Needles
Chemical Name or Material L(-)-Menthol, 99.7%
Show More Show Less
compliance-icons
Product Identifier
  • L(-)-Menthol
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P314-Get medical advice/attention if you feel unwell.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.