missing translation for 'onlineSavingsMsg'
Learn More

L-Methionine, Dr. Ehrenstorfer

Product Code. 30313330
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30313330 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313330 Supplier Dr. Ehrenstorfer Supplier No. DREC15021200
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Methionine
CAS 63-68-3
Analyte or Component Names L-Methionine
Shipping Condition Room Temperature
Molecular Formula C5 H11 N O2 S
Certifications/Compliance ISO/IEC 17025
Synonym (2S)-2-Amino-4-(methylsulfanyl)butanoic acid, L-Methionine, Methionine, L- (8CI), (S)-2-Amino-4-(methylthio)butanoic acid, 2-Amino-4-(methylthio)butyric acid, Acimethin, Butanoic acid, 2-amino-4-(methylthio)-, (S)-, Cymethion, L-Homocysteine, S-methyl-, L-α-Amino-γ-methylthiobutyric acid, Meth-o-Tas, Methionine, Methiorep, NSC 22946, S-Methionine, S-Methyl-L-homocysteine, Secretory peptide (human clone HTXDU73), h-Met-oh, l-Methionine, α-Amino-γ-methylmercaptobutyric acid, γ-Methylthio-α-aminobutyric acid, Phenylalanine Impurity B
InChI Formula InChI=1S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-/m0/s1
SMILES CSCC[C@H](N)C(=O)O
IUPAC Name (2S)-2-amino-4-methylsulfanylbutanoic acid
Molecular Weight (g/mol) 149.21
Quantity 100 mg
Formula Weight 149.051 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.