missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Phenylephrine hydrochloride, 99%

Product Code. 10432211 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10432211 10 g Glass bottle 10g
10032700 25 g Glass bottle 25g
10249493 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10432211 Supplier Thermo Scientific Acros Supplier No. 207240100
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 61-76-7 | C9H14ClNO2 | 203.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-76-7
Molecular Formula C9H14ClNO2
Molecular Weight (g/mol) 203.67
MDL Number MFCD00012605,MFCD00044749
InChI Key OCYSGIYOVXAGKQ-UHFFFAOYNA-N
Synonym phenylephrine hydrochloride, phenylephrine hcl, l-phenylephrine hydrochloride, metaoxedrine chloride, neosympatol, synethenate, almefrin, consdrin, emagrin, fenilfar
PubChem CID 5284443
ChEBI CHEBI:8094
IUPAC Name 3-[(1R)-1-hydroxy-2-(methylamino)ethyl]phenol;hydrochloride
SMILES [H+].[Cl-].CNCC(O)C1=CC=CC(O)=C1

Specifications

Melting Point 140°C to 145°C
Color White
Loss on Drying 1% max. (1 g, 105°C)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HOC6H4CH(CH2NHCH3)OH·HCl
Quantity 10 g
Merck Index 15, 7398
Specific Rotation Condition −47° (20°C c=2,H2O)
Specific Rotation -47
Formula Weight 203.67
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material L(-)-Phenylephrine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • L(-)-Phenylephrine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.