missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Selectin/CD62L, Rabbit anti-Mouse, Clone: MEL-14, Novus Biologicals™
Rabbit Monoclonal Antibody
275.00 € - 595.00 €
Specifications
| Antigen | L-Selectin/CD62L |
|---|---|
| Clone | MEL-14 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Block/Neutralize |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Blocking Assay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18352511
|
Novus Biologicals
NBP2-81083-0.025MG |
0.025 mg |
275.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18347550
|
Novus Biologicals
NBP2-81083-0.2MG |
0.2 mg |
595.00 €
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
L-Selectin/CD62L Monoclonal specifically detects L-Selectin/CD62L in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Block/Neutralize.Specifications
| L-Selectin/CD62L | |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Blocking Assay | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Immunology, Lipid and Metabolism, Myeloid derived Suppressor Cell, Signal Transduction | |
| PBS | |
| 6402 | |
| C3H/eb mouse B lymphoma 38C-13. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| MEL-14 | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Block/Neutralize | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM-1, LAM1LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L | |
| SELL | |
| IgG κ | |
| Protein A purified | |
| This antibody binds to mouse L-Selectin/CD62L the mouse peripheral lymph node homing receptor, which is a lectin-like cell surface protein rapidly down-regulated upon cell activation with phorbol 12-myristate 13-acetate. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title