missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Tartaric acid diammonium salt, 99%

Product Code. 10062710 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10062710 5 g Glass bottle 5g
10512041 250 g Glass bottle 250g
10599063 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10062710 Supplier Thermo Scientific Acros Supplier No. 211810050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3164-29-2 | C4H12N2O6 | 184.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3164-29-2
Molecular Formula C4H12N2O6
Molecular Weight (g/mol) 184.15
MDL Number MFCD00013073
InChI Key NGPGDYLVALNKEG-OLXYHTOASA-N
Synonym ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate
PubChem CID 13652278
ChEBI CHEBI:63075
SMILES N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O

Specifications

Color White
Density 1.6000g/mL
pH 6.0 to 7.0 (5% soln.)
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Glass bottle
Linear Formula [-CH(OH)CO2NH4]2
Quantity 5 g
Merck Index 09, 591
Specific Rotation Condition + 32.40 (20.00°C c=1.84,H2O)
Specific Rotation +31.4° to +33.4° (20°C, 589nm) (c=1.84, H2O)
IUPAC Name diazanium;(2R,3R)-2,3-dihydroxybutanedioate
Formula Weight 184.15
Percent Purity ≥98.5%
Insoluble Matter 0.005% max.
Physical Form Crystals
Chemical Name or Material L(+)-Tartaric acid diammonium salt, 0.99
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.