missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Tartaric acid diammonium salt, 99%

Product Code. 10512041 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10512041 250 g Glass bottle 250g
10062710 5 g Glass bottle 5g
10599063 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10512041 Supplier Thermo Scientific Acros Supplier No. 211812500
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
In Stock
Add to basket
Denna artikel kan inte returneras. Se returpolicy

CAS: 3164-29-2 | C4H12N2O6 | 184.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 3164-29-2
Molecular Formula C4H12N2O6
Molecular Weight (g/mol) 184.15
MDL Number MFCD00013073
InChI Key NGPGDYLVALNKEG-OLXYHTOASA-N
Synonym ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate
PubChem CID 13652278
ChEBI CHEBI:63075
SMILES N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O

Specifikationer

Color White
Density 1.6000g/mL
pH 6.0 to 7.0 (5% soln.)
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Glass bottle
Linear Formula [-CH(OH)CO2NH4]2
Quantity 250 g
Merck Index 09, 591
Specific Rotation Condition + 32.40 (20.00°C c=1.84,H2O)
Specific Rotation +31.4° to +33.4° (20°C, 589nm) (c=1.84, H2O)
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid diamine
Formula Weight 184.15
Percent Purity ≥98.5%
Insoluble Matter 0.005% max.
Physical Form Crystals
Chemical Name or Material L(+)-Tartaric acid diammonium salt, 0.99
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.