missing translation for 'onlineSavingsMsg'
Learn More

L-Theanine, MedChemExpress

Product Code. 16456636
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
Unit Size:
100mg
200mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16456636 100 mg 100mg
16411166 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16456636 Supplier Medchem Express Supplier No. HY15121
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L-Theanine (L-Glutamic Acid γ-ethyl amide)is a non-protein amino acid contained in green tea leaves, which blocks the binding of L-glutamic acid to glutamate receptors in the brain, and with neuroprotective and anti-oxidative activities. L-Theanine causes anti-stress effects via the inhibition of cortical neuron excitation by oral intake.

L-Theanine (L-Glutamic Acid γ-ethyl amide)is a non-protein amino acid contained in green tea leaves, which blocks the binding of L-glutamic acid to glutamate receptors in the brain, and with neuroprotective and anti-oxidative activities. L-Theanine causes anti-stress effects via the inhibition of cortical neuron excitation by oral intake.

Specifications

Chemical Name or Material L-Theanine
Physical Form Solid
CAS 3081-61-6
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C7H14N2O3
For Use With (Application) Neuroscience-Neurodegeneration
Synonym L-Glutamic Acid γ-ethyl amide Nγ-Ethyl-L-glutamine
Solubility Information H2O : 150 mg/mL (861.08 mM; Need ultrasonic)
SMILES OC([C@@H](N)CCC(NCC)=O)=O
Molecular Weight (g/mol) 174.2
Formula Weight 174.2
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.