missing translation for 'onlineSavingsMsg'
Learn More

L-Theanine, TRC

Product Code. 30132595
Change view
Click to view available options
Quantity:
50 mg
250 mg
2.5 g
Unit Size:
2.5g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132595 2.5 g 2.5g
30007808 250 mg 250mg
30003851 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132595 Supplier Toronto Research Chemicals Supplier No. TRCT3405002.5G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3081-61-6
Molecular Formula C7 H14 N2 O3
Molecular Weight (g/mol) 174.2
Synonym L-Glutamine, N-ethyl-, Glutamine, N-ethyl-, L- (6CI,7CI,8CI), L-Theanine, N-Ethyl-L-glutamine, NSC 21308, Nγ-Ethyl-L-glutamine, Suntheanine, Theanin, Theanine
IUPAC Name (2S)-2-amino-5-(ethylamino)-5-oxopentanoic acid
SMILES CCNC(=O)CC[C@H](N)C(=O)O

Specifications

InChI Formula InChI=1S/C7H14N2O3/c1-2-9-6(10)4-3-5(8)7(11)12/h5H,2-4,8H2,1H3,(H,9,10)(H,11,12)/t5-/m0/s1
Quantity 2.5 g
Formula Weight 174.1004
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material L-Theanine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.