accessibility menu, dialog, popup

UserName

H3CHNONH2OHO

CAS RN 3081-61-6

IUPAC Name: 2-amino-4-(ethylcarbamoyl)butanoic acid
Synonyms: theanine
Molecular Weight (g/mol): 174.2
Molecular Formula: C7H14N2O3
InChi Key: DATAGRPVKZEWHA-UHFFFAOYNA-N
SMILES: CCNC(=O)CCC(N)C(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)