Learn More
L1CAM Monoclonal Antibody (OTI1A8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA501437S
Encompass_Preferred
 View more versions of this product
Product Code. 17564241
  • 242.00 € / 30µL
Description
L1CAM Monoclonal antibody specifically detects L1CAM in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications
Specifications
L1CAM
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P32004
Mouse
Affinity chromatography
RUO
3897
-20° C, Avoid Freeze/Thaw Cycles
IgG2a
Flow Cytometry, Immunocytochemistry, Western Blot
OTI1A8
Unconjugated
L1CAM
antigen identified by monoclonal antibody R1; CAML1; CD 171; CD171; CD171 molecule; HSAS; HSAS1; Hyd; L1; L1 cell adhesion molecule; L1CAM; MASA; MIC5; N-CAM L1; NCAML1; N-CAML1; N-CAM-L1; NCAM-L1; Nerve-growth factor-inducible large external glycoprotein; neural cell adhesion molecule L1; NILE; S10; sCD171; sL1 CAM; sL1CAM; soluble CD 171; soluble CD171; soluble L1 CAM; soluble L1CAM; SPG1
Full length human recombinant protein of human L1CAM produced in HEK293T cell.
30 μL