Learn More
La Crosse Virus Glycoprotein 1 Antibody (807.31), DyLight 680 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-20121FR
Product Code. 18584539
  • 482.00 € / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
La Crosse Virus Glycoprotein 1 Monoclonal antibody specifically detects La Crosse Virus Glycoprotein 1 in Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
La Crosse Virus Glycoprotein 1
Monoclonal
DyLight 680
Bunyavirus La Crosse Glycoprotein 1, G1 envelope glycoprotein, Glycoprotein C, La Crosse Virus Gc, LCV G1, M polyprotein G1 chain
This antibody was raised by immunising BALB/c mice with La Crosse viruses and the harvested spleens were fused with mouse P3X63Ag8.653 myeloma cells.
0.1 mL
Primary
Virus
Purified
Western Blot, Flow Cytometry, ELISA
807.31
50mM Sodium Borate
Mouse
Protein A purified
RUO
956555
Store at 4°C in the dark.
IgG1 κ