Learn More
Calprtectin-Monocyte/Macrophage/L1 protein Ab-1 Mouse Monoclonal Antibody, Epredia™
Epredia™ Calprtectin-Monocyte/Macrophage/L1 protein Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
Brand: Epredia MS-148-PABX
Description
Host Species: Mouse
Clone: MAC 387
Isotype: IgG1
Species Reactivity: Human, Baboon, Monkey, Pig, Cat, Dog, Guinea pig, Rat, and Mouse. Others not known.
Epitope: Not determined
Immunogen: Affinity purified monocyte membrane preparation
Positive Control: Tonsil, lymph node, or spleen
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| Macrophage/L1 Protein/Calprotectin Ab-1 | |
| Monoclonal | |
| 1mg/mL | |
| Mouse | |
| 200 μL | |
| Hematopoietic | |
| Canine, Feline, Guinea Pig, Human, Mouse, Non-human Primate, Pig, Rat | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| MAC 387 | |
| Unconjugated | |
| Affinity purified monocyte membrane preparation | |
| IVD | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.