missing translation for 'onlineSavingsMsg'
Learn More

CD117/c-Kit/SCF-Receptor Ab-3 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments.

Brand:  Epredia MS-289-P1

 View more versions of this product

Product Code. 12680827

  • ONLINE EXCLUSIVE PRICE
    444.00 €
    372.90 € / 500µL
    Save 71.10 €
    16% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

c-kit is a transmembrane receptor tyrosine kinase. It binds to stem cell factor (SCF). It is expressed in many tissues and cells. C-kit/SCf is involved in development of several lineages of stem cells, such as germ cells, neural crest-derived melanocytes and hematopoietic precursor cells. Ab-3 does not interfere with the binding of SCF to c-kit

Host Species: Mouse

Clone: K45

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Extracellular second N-terminus Ig-like domain of c-kit

Immunogen: Recombinant human extracellular c-kit fragment

Molecular Weight: 145kDa

Positive Control: Skin, GI Stromal Tumor (GIST)

Cellular Localization: Membrane/cytoplasmic

Recommended for:

  • Immunohistochemistry (Frozen or fixed in methacarn or Stieve's and embedded in paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD117/c-Kit/SCF-Receptor Ab-3
Monoclonal
200μg/mL
Mouse
500 μL
Primary
Concentrated
Immunofluorescence, Immunohistochemistry (Frozen)
K45
Unconjugated
Recombinant human extracellular c-kit fragment
RUO
Human
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.