missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD138 Ab-2, Mouse Monoclonal Antibody

Obtain accurate and reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

Brand:  Epredia™ MS-1793-S0

 View more versions of this product

Product Code. 12660157

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

CD138/Syndecan-1 is a transmembrane heparin sulphate proteoglycan which is made up of one core protein and five glycosaminoglycan. CD138 is expected to play a role in cell adhesion. It is expressed on the surface of pre-B cells and plasma cells but is absent from mature B cells. It is a selective marker for B cell lymphoblastic leukemia and lymphoplasmocytoid leukemia. It is lost from the apoptotic myeloma cells; hence, it is a useful marker for viable myeloma cells.

Host Species: Mouse

Clone: MI15

Isotype: IgG1/κ

Species Reactivity: Human. Others not known

Epitope: Ectodomain

Immunogen: U-266 cells

Molecular Weight: 30.5kDa (core protein); 85-92kDa (depending upon glycosaminoglycans)

Positive Control: U-266 cells, Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD138 Ab-2
Monoclonal
Unconjugated
U-266 cells
100 μL
Cell Adhesion
Human
IgG2a κ
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
MI15
Mouse
U-266 cells, tonsil
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.