missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD23 Ab-1, Mouse Monoclonal Antibody

Ensure accurate and reproducible results in immunohistochemistry experiments with Epredia™ CD23 Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-729-S1

 View more versions of this product

Product Code. 12602187

  • ONLINE EXCLUSIVE PRICE
    614.00 €
    509.31 € / 500µL
    Save 104.69 €
    17% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD23 is identical to low affinity IgE receptor found on B cells. CD23 is expressed on a subpopulation of peripheral blood cells, B lymphocytes and on EBV transformed B lymphoblastoid cell lines. CD23 is also detected in neoplastic cells from cases of B cell chronic lymphocytic leukemia and some cases on centroblastic/centrocytic lymphoma.

Host Species: Mouse

Clone: 1B12

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat. Others not tested.

Epitope: Extracellular

Immunogen: Recombinant external domain of CD23 protein

Molecular Weight: 45 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD23 Ab-1
Monoclonal
Unconjugated
Recombinant external domain of CD23 protein
IVD
Primary
Concentrated
Immunohistochemistry (Paraffin)
1B12
Mouse
500 μL
Oncogenesis
Human
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.