Learn More
Epredia™ Lab Vision™ CD23 Ab-1, Mouse Monoclonal Antibody
Ensure accurate and reproducible results in immunohistochemistry experiments with Epredia™ CD23 Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-729-RQ
Description
CD23 is identical to low affinity IgE receptor found on B cells. CD23 is expressed on a subpopulation of peripheral blood cells, B lymphocytes and on EBV transformed B lymphoblastoid cell lines. CD23 is also detected in neoplastic cells from cases of B cell chronic lymphocytic leukemia and some cases on centroblastic/centrocytic lymphoma.Host Species: Mouse
Clone: 1B12
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with rat. Others not tested.
Epitope: Extracellular
Immunogen: Recombinant external domain of CD23 protein
Molecular Weight: 45 kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| CD23 Ab-1 | |
| Monoclonal | |
| Unconjugated | |
| Recombinant external domain of CD23 protein | |
| IVD | |
| Primary |
| Immunohistochemistry (Paraffin) | |
| 1B12 | |
| Mouse | |
| 12 mL | |
| Oncogenesis | |
| Ready-to-Use |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.