missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD3 (Early T-Cell Marker) Rabbit Monoclonal Antibody,

Stain normal and neoplastic T cells in formalin-fixed, paraffin-embedded tissues with Epredia™ CD3 (Early T-Cell Marker), Rabbit Monoclonal Antibody.

202.00 € - 1020.00 €

Specifications

Antigen CD3
Clone SP7
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Quantity Form Price Quantity & Availability  
Product Code Brand Quantity Form Price Quantity & Availability  
12604737
View Documents
Epredia™
RM-9107-S0
100 μL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12644527
View Documents
Epredia™
RM-9107-S1
500 μL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12643707
View Documents
Epredia™
RM-9107-S
1 mL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12623707
View Documents
Epredia™
RM-9107-R7
7 mL Prediluted
ONLINE EXCLUSIVE PRICE
475.00 €
435.05 € / 7mL
Save 39.95 €
8% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12633707
View Documents
Epredia™
RM-9107-RQ
12 mL Ready-to-Use This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This antibody reacts with the intracytoplasmic portion of the CD3 antigen expressed by T cells. It stains human T cells in both the cortex and medulla of the thymus and in peripheral lymphoid tissues. This antibody is suitable for staining normal and neoplastic T cells in formalin-fixed, paraffin-embedded tissues.

Host Species: Rabbit

Clone: SP7

Isotype: IgG

Species Reactivity: Human, Baboon, Monkey, Horse, Dog and Cat. Others not tested.

Epitope: 156-168aa

Immunogen: A synthetic 13-mer peptide corresponding to aa 156-168 of the ε-chain of human CD3 protein

Positive Control: Jurkat cells, Tonsil

Cellular Localization: Membrane

Clone SP7 is excellent for staining of formal/paraffin.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

CD3
Immunohistochemistry (Paraffin)
Unconjugated
IVD
A synthetic 13-mer peptide corresponding to aa 156-168 of the epsilon-chain of human CD3 protein
SP7
Monoclonal
Rabbit
Cancer and Tumor Biology
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.