missing translation for 'onlineSavingsMsg'
Learn More

CD31/PECAM-1 (Endothelial Cell Marker) Rabbit Polyclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry procedures with Epredia™ CD31/PECAM-1 (Endothelial Cell Marker), Rabbit Polyclonal Antibody.

Brand:  Epredia RB-10333-P

 View more versions of this product

Product Code. 12642787

  • ONLINE EXCLUSIVE PRICE
    728.00 €
    657.57 € / 1mL
    Save 70.43 €
    9% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD31 is a glycoprotein expressed on endothelial cells and in platelets. It is known to be involved in cell signaling and cell adhesion. Antibody to CD31 is of value in the study of benign and malignant vascular tumors. Staining for CD31 has also been used to measure angiogenesis, which reportedly predicts tumor recurrence.

Host Species: Rabbit

Species Reactivity: Human. Others not tested.

Epitope: C-terminal

Immunogen: Synthetic peptide corresponding to C-terminus of mouse CD31 protein

Molecular Weight: 100 and 130 kDa

Positive Control: Tonsil, Placenta

Cellular Localization: Endothelial cells in blood vessel of tonsil

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD31 (Endothelial Cell Marker)
Polyclonal
200µg/mL
Rabbit
Tonsil
RUO
Primary
Concentrated
Immunohistochemistry (Paraffin)
Polyclonal
Unconjugated
Synthetic peptide corresponding to C-terminus of mouse CD31 protein
1 mL
Cancer and Tumor Biology
Human
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.