missing translation for 'onlineSavingsMsg'
Learn More

CD68/Macrophage Marker Ab-3 Mouse Monoclonal Antibody, Epredia™

Product Code. 12651317 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Supernatant
Concentration:
1mg/mL
200μg/mL
Unit Size:
0.1mL
0.2mL
100µL
12mL
1mL
500µL
7mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Form Concentration unitSize
12651317 500 μL Concentrated 200μg/mL 500µL
12641317 100 μL Concentrated 200μg/mL 100µL
12691317 100 μL Concentrated 1mg/mL 0.1mL
12681317 200 μL Concentrated 1mg/mL 0.2mL
12621317 1 mL Concentrated 200μg/mL 1mL
12601327 7 mL Prediluted - 7mL
12611327 12 mL Supernatant - 12mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 12651317 Supplier Epredia Supplier No. MS397P1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD68/Macrophage Marker Ab-3, Mouse Monoclonal Antibody.

CD68 is expressed on macrophages and monocytes.

Host Species: Mouse

Clone: KP1

Isotype: IgG1/κ

Species Reactivity: Human, Monkey, Cat and Rat. Does not react with pig, dog and chicken.

Epitope: Not determined

Immunogen: Subcellular fraction of human alveolar macrophages

Molecular Weight: 110 kDa

Positive Control: Tonsil, lymph node, or spleen

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD68/Macrophage Marker Ab-3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone KP1
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Subcellular fraction of human alveolar macrophages
Quantity 500 μL
Regulatory Status IVD
Research Discipline Hematopoietic
Primary or Secondary Primary
Target Species Feline, Human, Non-human Primate, Rat
Form Concentrated
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.