missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD71/Transferrin Receptor Ab-3, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry applications with Epredia™ CD71/Transferrin Receptor Ab-3, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1096-S

 View more versions of this product

Product Code. 12658936

  • ONLINE EXCLUSIVE PRICE
    655.00 €
    539.99 € / 1mL
    Save 115.01 €
    17% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Cell surface transferrin receptor (CD71) is a disulfide-bonded homodimeric glycoprotein. Ligand for transferrin receptor is the serum iron transport protein, transferrin. This receptor is broadly distributed in carcinomas, sarcomas, leukemias, and lymphomas. Transferrin receptor (CD71) has been reported to be associated with cell proliferation in both normal and neoplastic tissues and useful in predicting clinical behavior or response to therapy in a number of malignancies including breast cancer.

Host Species: Mouse

Clone: 10F11

Isotype: IgG2b

Species Reactivity: Human. Others not known.

Epitope: Not determined

Molecular Weight: ∽90-95 kDa

Positive Control: Placenta

Cellular Localization: Granular cytoplasmic and membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD71/Transferrin Receptor Ab-3
Monoclonal
Unconjugated
Tonsil
RUO
Human
IgG2b
Immunohistochemistry (Paraffin)
10F11
Mouse
1 mL
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.