Learn More
CDC25C Ab-1 Mouse Monoclonal Antibody, Epredia™
CDC25C Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
Brand: Epredia MS-751-P0
Description
Cyclin-dependent kinases can be activated by CDC25, which removes inhibitory phosphates from tyrosine and threonine residues. At least three CDC25 genes (CDC25A, CDC25B, and CDC25C) have been identified in humans. Whereas CDC25A and CDC25B are expressed throughout the cell cycle, with peak expression in G1 for CDC25A and in both G1-S-phase and G2 for CDC25B, CDC25C is predominantly expressed in G2.Host Species: Mouse
Clone: 25C14; same as TC-14
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with mouse and rat. Others not known.
Epitope: aa 1-150
Immunogen: Recombinant human CDC25C protein
Molecular Weight: 53-56 kDa
Positive Control: HeLa, HT29 or LS174T cells
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Not Suitable)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Denatured verified)
Note:
USA: RUO; Int'l: RUO
Specifications
| CDC25C Ab-1 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 100 μL | |
| Cell Cycle | |
| Human | |
| IgG2a κ |
| Immunofluorescence, Immunoprecipitation, Western Blot | |
| 25C14; same as TC-14 | |
| Unconjugated | |
| Recombinant human CDC25C protein | |
| RUO | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.