Learn More
Collagen II Ab-1 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in western blotting and immunofluorescence experiments with Epredia™ Collagen II Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia MS-234-PABX
Description
Host Species: Mouse
Clone: 5B2.5
Isotype: IgG2a/κ
Species Reactivity: Human, Chicken and Mouse. Others not known.
Epitope: GGFDEK, slightly preceding the lysine residue involved in crosslinking. Two terminal glycine residues constitute a key structural element of the epitope. The epitope is lost after digestion of lathyritic type II collagen with recombinant stromelysin.
Immunogen: A purified preparation of lathyritic type II collagen from embryonic chicken sternum
Positive Control: Cartilage
Cellular Localization: Cartilage matrix
Recommended for:
- Immunohistochemistry (Not suitable)
- Western Blotting
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
| Collagen II Ab-1 | |
| Monoclonal | |
| 1mg/mL | |
| Mouse | |
| 200 μL | |
| Extracellular Matrix | |
| Chicken, Human, Mouse | |
| IgG2a κ |
| Immunofluorescence, Western Blot | |
| 5B2.5 | |
| Unconjugated | |
| A purified preparation of lathyritic type II collagen from embryonic chicken sternum | |
| RUO | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.