Learn More
Epredia™ Lab Vision™ DP-1 Transcription Factor Ab-6 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Ensure accurate, reproducible results in western blotting experiments with Epredia™ DP-1 Transcription Factor Ab-6, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-1056-P
Description
DP family contains at least two members, DP-1 and -2. Structurally, DP-1 and -2 are relatively conserved, yet their expression appears to be tissue specific. E2F-1, a functional target for the tumor suppressor protein Rb, heterodimers with DP-1 and forms an active E2F transcriptional complex. The interaction between members of the E2F and the DP families is mediated, in part, by a leucine repeat which is weakly conserved between the two families.Host Species: Mouse
Clone: 1DP06 (TFD10)
Isotype: IgG1/κ
Species Reactivity: Human, Mouse and Rat. Others not known.
Epitope: aa 83-204
Immunogen: Recombinant DP-1 protein
Molecular Weight: 45 kDa
Positive Control: MAD109 or Raji cells, Tonsil
Cellular Localization: Cytoplasm and nucleus
Recommended for:
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
| DP-1 Transcription Factor Ab-6 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 1 mL | |
| Transcription Regulation | |
| Human, Mouse, Rat | |
| IgG2a κ |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1DP06; same as TFD10 | |
| Unconjugated | |
| Recombinant DP-1 protein | |
| RUO | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.