Learn More
Epredia™ Lab Vision™ Epstein-Barr Virus/LMP Ab-1 Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epstein-Barr Virus/LMP Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-1458-S
Description
Host Species: Mouse
Clone: CS1 + CS2 + CS3 + CS4
Isotype: IgG1/κ
Species Reactivity: EBV-positive cells
Epitope: Located in the internal part of the membrane-associated viral LMP
Immunogen: EBV-encoded recombinant latent membrane protein
Molecular Weight: 60 kDa (LMP)
Positive Control: EBV-positive lymphoblastoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| Epstein-Barr Virus/LMP Ab-1 | |
| Monoclonal | |
| Unconjugated | |
| Mouse | |
| 1 mL | |
| Infectious Disease | |
| Ebola virus | |
| Concentrated |
| Immunohistochemistry (Paraffin) | |
| CS1+CS2+CS3+CS4 | |
| 1:25-1:50 for 60 min at RT | |
| EBV-encoded recombinant latent membrane protein | |
| ASR, IVD | |
| Primary | |
| Store vial at 4deg.C. When stored at 2 to 8deg.C, | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.