missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Factor XIII A Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Factor XIII A Ab-1, Mouse Monoclonal Antibody.

158.10 € - 647.70 €

Specifications

Antigen Factor XIII A Ab-1
Clone AC-1A1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 6
Product Code Brand Quantity Concentration Price Quantity & Availability  
Product Code Brand Quantity Concentration Price Quantity & Availability  
12659346
View Documents
Epredia™
MS-1237-P0
100 μL 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12669346
View Documents
Epredia™
MS-1237-P1
500 μL 200μg/mL
ONLINE EXCLUSIVE PRICE
462.00 €
387.79 € / 500µL
Save 74.21 €
16% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12649346
View Documents
Epredia™
MS-1237-P
1 mL 200μg/mL
ONLINE EXCLUSIVE PRICE
725.00 €
599.25 € / 1mL
Save 125.75 €
17% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12679346
View Documents
Epredia™
MS-1237-P1ABX
100 μL 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12689346
View Documents
Epredia™
MS-1237-PABX
200 μL 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12629356
View Documents
Epredia™
MS-1237-R7
7 mL
ONLINE EXCLUSIVE PRICE
408.00 €
341.01 € / 7mL
Save 66.99 €
16% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Detect factor XIII in both reduced and non-reduced forms. This abtibody does not react with human Factor XIII B-chain or human Factor XII. Factor XIII is a Β-globulin found in plasma and is composed of two subunits. Factor XIII-A is the catalytic subunit and is a dimer of M.W. 160 kDa. Factor XIII is present in plasma as an alpha2Β2 heterodimer (M.W. 320 kDa); whereas in platelets, only the α2 unit exists. Factor XIIIa is a dermal dendrocyte marker and shows variable reaction with these types of tumors. It can be used for histiocytic phenotyping and has been reported to mark capillary hemangiomas and tumors of the central nervous system. Factor XIII has also been used with CD34 to differentiate between dermatofibroma and dermatofibrosarcoma protuberans.

Host Species: Mouse

Clone: AC-1A1

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant protein corresponding to A-subunit of coagulation Factor XIII

Molecular Weight: 160 kDa (unreduced dimmer); 80 kDa (reduced monomer)

Positive Control: Recombinant human Factor XIII A-subunit; Capillary hemangioma, dermatofibroma and placenta

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
Specifications

Specifications

Factor XIII A Ab-1
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
IVD
Recombinant protein corresponding to A-subunit of coagulation Factor XIII
AC-1A1
Monoclonal
Mouse
Hematopoietic
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.