missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Hepatitis B Virus Surface Antigen (HBVsAg) Ab-1 Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Hepatitis B Virus Surface Antigen (HBVsAg) Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-314-S1

 View more versions of this product

Product Code. 12630967

  • ONLINE EXCLUSIVE PRICE
    326.00 €
    272.55 € / 500µL
    Save 53.45 €
    16% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Hepatitis B virus causes type B hepatitis and it belongs to hepadnavirus family. It is spherical in shape with a diameter of 42nm. It contains a 27nm partially double stranded DNA core enclosed within a lipoprotein coat. The antigenic activity of the nucleocapsid core is designated as hepatitis B core antigen. The antigens in the outer surface are called as hepatitis B virus surface antigens. Core antigens are localized within the nuclei whereas the surface antigens are present in the cytoplasm of the infected cells. Antibodies to surface antigens appear in circulation at an early stage of infection whereas the antibodies to the core antigens are detected after several weeks.

Host Species: Mouse

Clone: T9; same as 3E7

Isotype: IgG1/λ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified HBVsAg, subtype ayw3

Positive Control: Hepatitis B virus infected liver

Cellular Localization: Cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hepatitis B Virus Surface Antigen (HBVsAg) Ab-1
Monoclonal
Unconjugated
Purified HbsAg, subtype ayw3
ASR
Primary
Concentrated
Immunohistochemistry (Paraffin)
T9; same as 3E7
Mouse
500 μL
Infectious Disease
Human
IgG1 λ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.