missing translation for 'onlineSavingsMsg'
Learn More

L1 Cell Adhesion Molecule Ab-1 Mouse Monoclonal Antibody, Epredia™

Recommended for Immunohistochemistry (Formalin/paraffin), Western Blotting and Immunoprecipitation (Native and denatured), Epredia™ L1 Cell Adhesion Molecule Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia MS-770-P1

 View more versions of this product

Product Code. 12692287

  • ONLINE EXCLUSIVE PRICE
    487.00 €
    410.84 € / 500µL
    Save 76.16 €
    15% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The L1CAM gene, which is located in Xq28, is involved in three distinct conditions: 1) HSAS (hydrocephalus-stenosis of the aqueduct of Sylvius); 2) MASA (mental retardation, aphasia, shuffling gait, adductus thumbs); and 3) SPG1 (spastic paraplegia). The L1, neural cell adhesion molecule (L1CAM) also plays an important role in axon growth, fasciculation, neural migration and in mediating neuronal differentiation. Expression of L1 protein is restricted to tissues arising from neuroectoderm.

Host Species: Mouse

Clone: UJ127

Isotype: IgG1/k

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Homogenous suspension of 16-week human fetal brain

Molecular Weight: 220-240kDa

Positive Control: IMR-5 cells Neuroblastoma

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

L1 Cell Adhesion Molecule Ab-1
Monoclonal
200μg/mL
Mouse
500 μL
Cancer and Tumor Biology
Human
IgG2a κ
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
UJ127
Unconjugated
Homogenous suspension of 16 week human fetal brain
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.