missing translation for 'onlineSavingsMsg'
Learn More

MMP-3 (Stromelysin-1) Ab-1 Mouse Monoclonal Antibody, Epredia™

Epredia™ MMP-3 (Stromelysin-1) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

170.00 € - 740.00 €

Specifications

Antigen MMP-3 (Stromelysin-1) Ab-1
Clone SL-1 ID3
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Form Concentrated
View More Specs

Products 6
Product Code Brand Quantity Conjugate Concentration Price Quantity & Availability  
Product Code Brand Quantity Conjugate Concentration Price Quantity & Availability  
12602367
View Documents
Epredia
MS-809-B0
100 μL Biotin 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12612367
View Documents
Epredia
MS-809-B1
500 μL Biotin 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12632367
View Documents
Epredia
MS-809-P0
100 μL Unconjugated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12642367
View Documents
Epredia
MS-809-P1
500 μL Unconjugated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12652367
View Documents
Epredia
MS-809-P1ABX
100 μL Unconjugated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12662367
View Documents
Epredia
MS-809-PABX
200 μL Unconjugated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Matrix metalloproteinases (MMP) are proteolytic enzymes capable of degrading connective tissue components. Imbalanced secretion of certain MMP or disturbances in the differential control of MMP by tissue inhibitor of MMPs (TIMPs) are implicated in the invasive potential of malignant tumors. Stromelysin (MMP3/Transin) degrades several matrix components including the core proteins of proteoglycans, laminin and non-helical regions of collagens.

Host Species: Mouse

Clone: SL-1 ID3

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: APMA (4-Aminophenylmercuric acetate) activated Human stromelysin-1 (SL-1)

Molecular Weight: 54/59kDa (proform) and 44/49kDa (active form)

Positive Control: Conditioned, serum-free medium from TPA-treated human endometrial cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Not Suitable)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

General References:

1. Lein M, et al. Urologe A 199837(4):377-381.

2. Clavel C, et al. Bull Cancer 199279:261-270.

3. Muller JC, et al. Biol Chem 1997378:1475-1480.

Specifications

Specifications

MMP-3 (Stromelysin-1) Ab-1
Immunofluorescence, Western Blot
Concentrated
RUO
Human
IgG2a κ
SL-1 ID3
Monoclonal
Mouse
Proteases And Inhibitors
APMA (4-Aminophenylmercuric acetate) activated Human stromelysin-1 (SL-1)
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.