Learn More
MMP-3 (Stromelysin-1) Ab-1 Mouse Monoclonal Antibody, Epredia™
Epredia™ MMP-3 (Stromelysin-1) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
Brand: Epredia MS-809-P0
Description
Matrix metalloproteinases (MMP) are proteolytic enzymes capable of degrading connective tissue components. Imbalanced secretion of certain MMP or disturbances in the differential control of MMP by tissue inhibitor of MMPs (TIMPs) are implicated in the invasive potential of malignant tumors. Stromelysin (MMP3/Transin) degrades several matrix components including the core proteins of proteoglycans, laminin and non-helical regions of collagens.Host Species: Mouse
Clone: SL-1 ID3
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: APMA (4-Aminophenylmercuric acetate) activated Human stromelysin-1 (SL-1)
Molecular Weight: 54/59kDa (proform) and 44/49kDa (active form)
Positive Control: Conditioned, serum-free medium from TPA-treated human endometrial cells
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Not Suitable)
- Western Blotting
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
| MMP-3 (Stromelysin-1) Ab-1 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 100 μL | |
| Proteases And Inhibitors | |
| Human | |
| IgG2a κ |
| Immunofluorescence, Western Blot | |
| SL-1 ID3 | |
| Unconjugated | |
| APMA (4-Aminophenylmercuric acetate) activated Human stromelysin-1 (SL-1) | |
| RUO | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.